missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebacoyl-L-Carnitine Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™
$245.63 - $1003.18
Specifications
| CAS | 102636-85-1 (Free Acid) |
|---|---|
| Molecular Formula | C17H31NO6 · xLi+ |
| Linear Formula | C17H31NO6 · xLi+ |
| Formula Weight | 345.43 (Free Acid Basis) |
| Percent Purity | ≥93% (HPLC) |
Specifications
| 102636-85-1 (Free Acid) | |
| C17H31NO6 · xLi+ | |
| ≥93% (HPLC) |
| C17H31NO6 · xLi+ | |
| 345.43 (Free Acid Basis) | |
| Analytical Standard |