missing translation for 'onlineSavingsMsg'
Learn More

Sebacic Acid, 98%

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol

$58.47 - $1,899.09

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004440
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC132631000
View Documents Promotion Details
N/A 100 g Plastic bottle
Each for $58.47
Only null left
 
AC132630010
View Documents Promotion Details
N/A 1 kg Plastic bottle
Each for $242.81
Only null left
 
AC132630025
View Documents Promotion Details
N/A 2.5 kg Plastic bottle
Each for $525.96
Only null left
 
AC132630100
View Documents Promotion Details
N/A 10 kg Plastic drum
Each for $1,899.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-20-6
202.25
CXMXRPHRNRROMY-UHFFFAOYSA-N
5192
decanedioic acid
C10H18O4
MFCD00004440
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
CHEBI:41865
C(CCCCC(=O)O)CCCC(=O)O

Specifications

111-20-6
White
220°C
97.5% min. (GC)
HO2C(CH2)8CO2H
02, IV, 2078
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
CXMXRPHRNRROMY-UHFFFAOYSA-N
decanedioic acid
5192
202.25
Powder or Granules
131.0°C to 134.0°C
294.4°C (100.0 mmHg)
Authentic
C10H18O4
MFCD00004440
15, 8552
Solubility in water: 1g/L (20°C). Other solubilities: 0.1% in ether (17°C), freely soluble in alcohol, esters and ketones, sparingly soluble in hydrocarbons, insoluble in ethyl ether and carbon tetrachloride
C(CCCCC(=O)O)CCCC(=O)O
202.25
CHEBI:41865
98%
Sebacic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-845-5

SDS
Documents

Documents

RUO – Research Use Only