missing translation for 'onlineSavingsMsg'
Learn More

Sebacic acid, 98+%

Catalog No. AAA1415830
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1415830 250 g
AAA1415822 100 g
AAA141580B 1000 g
AAA141580I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1415830 Supplier Thermo Scientific Chemicals Supplier No. A1415830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol

Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

Solubility
Slightly soluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004440
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O

Specifications

Melting Point 131°C to 134°C
Density 1.271
Boiling Point 295°C (100 mmHg)
Flash Point 220°C (428°F)
Linear Formula HO2C(CH2)8CO2H
Quantity 250 g
Beilstein 1210591
Merck Index 14,8415
Solubility Information Slightly soluble in water.
Formula Weight 202.25
Percent Purity ≥98%
Chemical Name or Material Sebacic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 203-845-5
RTECSNumber VS0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only