missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing

Reliable and flexible dialysis tubing with 12,000 MWCO for separating macromolecules in educational chemistry, biology, and pharmaceutical labs.

Supplier:  Fisher Science Education™ S25645D

 View more versions of this product

Catalog No. S25645D


Only null left
Explore available promotions

Description

Description

  • Material: Seamless cellulose
  • Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
  • Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
  • Permeability: High permeability to water and low molecular weight compounds
  • Pore Diameter: Approx. 4.8 nm
  • Formulation: Glycerin, water, and 1% sulfur
  • Storage: Store in an airtight container; rehumidify if dried out
  • Diameter: 45 mm (1.13 in.)
  • Length: 30 m (100 ft.)
  • Quantity: Sold individually (Each)
  • Educational Use Only: No Certificate of Analysis (C of A) provided

This product is an Educational Grade Chemical — no C of A is available. ‡ Average values.

Specifications

Specifications

Dialysis Tubing
6.4 mm
10 ft.
Glycerin, water and 1% sulfur
Useful in analytical chemistry, biology, and pharmaceutical fields
Seamless Cellulose
0.25 in.
3 m
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions