missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Science Education™ Seamless Cellulose Dialysis Tubing
Description
- Material: Seamless cellulose
- Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
- Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
- Permeability: High permeability to water and low molecular weight compounds
- Pore Diameter: Approx. 4.8 nm
- Formulation: Glycerin, water, and 1% sulfur
- Storage: Store in an airtight container; rehumidify if dried out
- Diameter: 45 mm (1.13 in.)
- Length: 30 m (100 ft.)
- Quantity: Sold individually (Each)
- Educational Use Only: No Certificate of Analysis (C of A) provided
This product is for educational use - no C of A is available.
Specifications
Specifications
| Product Type | Dialysis Tubing |
| Material | Seamless Cellulose |
| Diameter (Metric) | 20.4 mm |
| Diameter (English) | 0.8 in. |
| Length (English) | 10 ft. |
| Length (Metric) | 3 m |
| Formulation | Glycerin, water and 1% sulfur |
| Quantity | Each |
| For Use With (Application) | Useful in analytical chemistry, biology, and pharmaceutical fields |