missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing

Catalog No. S25645G
Change view
Click to view available options
Diameter (Metric):
6.4 mm
15.9 mm
20.4 mm
28.6 mm
32 mm
Length (Metric):
3 m
30 m
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Length (Metric)
S25645G 32 mm 30 m
S25645F 15.9 mm 30 m
S25645B 28.6 mm 3 m
S25645A 20.4 mm 3 m
S25645 15.9 mm 3 m
S25645D 6.4 mm 3 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S25645G Supplier Fisher Science Education™ Supplier No. S25645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reliable and flexible dialysis tubing with 12,000 MWCO for separating macromolecules in educational chemistry, biology, and pharmaceutical labs.

  • Material: Seamless cellulose
  • Molecular Weight Cut-Off (MWCO): Retains molecules ≥12,000
  • Da Applications: Ideal for educational settings in analytical chemistry, biology, and pharmaceutical studies
  • Permeability: High permeability to water and low molecular weight compounds
  • Pore Diameter: Approx. 4.8 nm
  • Formulation: Glycerin, water, and 1% sulfur
  • Storage: Store in an airtight container; rehumidify if dried out
  • Diameter: 45 mm (1.13 in.)
  • Length: 30 m (100 ft.)
  • Quantity: Sold individually (Each)
  • Educational Use Only: No Certificate of Analysis (C of A) provided

This product is for educational use - no C of A is available.

Specifications

Product Type Dialysis Tubing
Material Seamless Cellulose
Diameter (Metric) 32 mm
Diameter (English) 0.8 in.
Length (English) 100 ft.
Length (Metric) 30 m
Formulation Glycerin, water and 1% sulfur
Quantity Each
For Use With (Application) Useful in analytical chemistry, biology, and pharmaceutical fields