missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XFp Fuel Inhibitor Pack

Catalog No. 103270100
Change view
Click to view available options
Quantity:
6 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
103270100 6 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103270100 Supplier Agilent Technologies™ Supplier No. 103270100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seahorse XFp Fuel Inhibitor Pack containing six single-use pouches, each with etomoxir, UK5099, and BPTES. For use with Seahorse XF HS Mini and XFp analyzers.

  • Uses specific inhibitors in different combinations to measure the dependency, capacity, and flexibility of cells to oxidize three major mitochondrial fuels: glucose (pyruvate), glutamine (glutamate), and long-chain fatty acids in basal energetic state
  • Compounds are reconstituted directly in the assay medium, eliminating the need for DMSO or other solvents
  • Each lot is rigorously tested and validated with functional assays to deliver consistent and robust results, eliminating the costs associated with failed experiments
  • Agilent™ Seahorse Mito Fuel Flex Test report generator simplifies data processing and transforms raw data into organized, shareable summary reports

Specifications

For Use With (Application) Cell Function
For Use With (Equipment) Other Fluorescence Detection Systems
Product Type Reagent Kit
Form Lyophilized
Quantity 6 Assays