missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XFp Cell Mito Stress Test Kit

Catalog No. 103010100
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
103010100 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 103010100 Supplier Agilent Technologies™ Supplier No. 103010100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seahorse XFp Cell Mito Stress Test Kit containing six single-use pouches, each with oligomycin, FCCP, and rotenone/antimycin A. For use with Seahorse XF HS Mini and XFp Analyzers.

  • Assay measures basal respiration, ATP-linked respiration, H+ (proton) leak, maximal respiration, spare respiratory capacity, and non-mitochondrial respiration using modulators of cellular respiration
  • Compounds are reconstituted directly in the assay medium, eliminating the need for DMSO or other solvents
  • Each lot is rigorously tested and validated with functional assays to deliver consistent and robust results, eliminating the costs associated with failed experiments
  • Upload your assay result file to your Agilent™ Seahorse Analytics account to calculate and report parameters of the XF Cell Mito Stress Test, share with collaborators, or export data to Microsoft Excel or GraphPad Prism for other graphing or analysis needs

Specifications

For Use With (Application) Cell Function
For Use With (Equipment) Other Fluorescence Detection Systems
Product Type Reagent Kit
Form Lyophilized
Quantity 1 Kit