missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDR42E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553156 Supplier Invitrogen™ Supplier No. PA553156
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: FQPFLTRTEV YKTGVTHYFS LEKAKKELGY KAQPFDLQEA VEWFKAHGHG RSSGSRDSEC Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 78%.

Specifications

Antigen SDR42E1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDR42E1
Gene Accession No. Q8WUS8
Gene Alias 4632417N05Rik; AI117603; HSPC105; RGD1308481; SDR42E1; short chain dehydrogenase/reductase family 42E, member 1; short-chain dehydrogenase/reductase family 42E member 1
Gene Symbols SDR42E1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SDR42E1. Recombinant protein control fragment (Product #RP-91054).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93517
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less