Learn More
Thermo Scientific™ Screw Vial Convenience Kit
Catalog No. 03393G
Change view
Click to view available options
Volume (Metric):
4 mL
8 mL
12 mL
16 mL
Color:
Clear
Closure Type:
Screw Cap, Open Top
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Closure Type
03393G 4 mL Clear Screw Cap, Open Top
03393A 8 mL Clear Screw Cap, Open Top
03393B 12 mL Clear Screw Cap, Open Top
03393C 16 mL Clear Screw Cap, Open Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03393G Supplier Thermo Scientific™ Supplier No. B79902
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
Description Screw Vial Convenience Kit, Open top Cap
Color Clear
Material Glass
Volume (Metric) 4 mL
Includes 4mL clear glass vial, open-top cap
No. per Pack 100
Closure Type Screw Cap, Open Top