missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Screw Vial Caps and Seals
Description
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Specifications
Specifications
| Color (Seal) | Red/White |
| Material (Liner) | PTFE/Silicone |
| Color | Black |
| Material | PTFE, Silicone |
| Septum | Red PTFE/White Silicone |
| For Use With (Equipment) | Thermo Scientific Spectra System AS3500/AS3000/AS1000/Surveyor/Accela LC, Agilent 1090, PE Autosystem |