Learn More
MilliporeSigma™ Supelco™ Screw Top Mini Flask with Hole Cap and Septum
Heat resistant borosilicate glass flasks provide an inert seal when used with a hole cap and PTFE silicone septum (included) or a Mininert valve.
$428.37 - $428.37
Specifications
Color Clear
Packaging Quantity 12/Pk.
Material Borosilicate Glass
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
111009770
View Documents
MilliporeSigma™ Supelco™
64715
10 mL
Each for $428.37
Only null left
Description
Description
Excellent for liquid-liquid extraction of trihalomethanes from drinking water. Also useful for sample collection, solution preparations, derivatization reactions, extractions, and many other applications.
Specifications
Specifications
Clear
12/Pk.
Borosilicate Glass