Learn More
MilliporeSigma™ Supelco™ Screw Top Mini Flask with Hole Cap and Septum
Catalog No. 111009770
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
111009770 10 mL
1 options
Catalog No. 111009770 Supplier MilliporeSigma™ Supelco™ Supplier No. 64715
Heat resistant borosilicate glass flasks provide an inert seal when used with a hole cap and PTFE silicone septum (included) or a Mininert valve.
Excellent for liquid-liquid extraction of trihalomethanes from drinking water. Also useful for sample collection, solution preparations, derivatization reactions, extractions, and many other applications.
Specifications
Color Clear
Volume (Metric) 10 mL
Packaging Quantity 12/Pk.
Material Borosilicate Glass