missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Screw Cap for Externally Threaded Sample Tubes (T501 and T502)

Catalog No. 361030265
Change view
Click to view available options
Color:
Natural
Orange
Red
White
Includes:
Lip seal
Silicone washer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
361030265 Natural Lip seal
23045269 Orange Silicone washer
23045270 Red Silicone washer
23045271 White Silicone washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361030265 Supplier Simport™ Scientific Supplier No. T501N
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Screw Cap for Externally Threaded Sample Tubes (T501 and T502) features long skirt and super-fast thread design that allows it to be removed or sealed with single turn.

  • Made of polypropylene
  • Autoclavable
  • For externally threaded tubes
  • Tubes sold separately

Specifications

Includes Lip seal
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
No. per Pack 1000 Pk.
Sterility Non-sterile
For Use With (Equipment) T501 and T502 Series Sample Tube
Disposable Single-use