Learn More
MilliporeSigma™ Supelco™ Screw Cap Black Polypropylene
Screw caps are available as hole caps for use with sealing septa.
$121.00
Specifications
No. per Pack 100
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Material Price Quantity & Availability  
111011218
View Documents Promotion Details
MilliporeSigma™ Supelco™
27439
4 mL Vial, 13-425 Thread PTFE/Silicone, Black Polypropylene (Top Hat)
Each for $121.00
Only null left
Description
Description
  • Used for capping water samples in amber glass vial comprising of a polytetrafluoroethylene silicon septum for the analysis of chlorinated volatile organic compounds in water samples using headspace-solid-phase microextraction with gas chromatography (HS-SPME-GC).
  • Screw caps (large, opening in top of cap, no septum, thread 8-425) are available as hole caps for use with sealing septa.
  • Polypropylene caps can withstand a maximum temperature of 135°C.
Specifications
Specifications
100