missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCP1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527354

Catalog No. PIPA527354


Only null left

Description

Description

Recommended positive controls: U87-MG, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Major component of the transverse filaments of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase.
Specifications

Chemical Identifiers

SCP1
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
SYCP1
A coiled-coil related protein specific for synapsed regions of meiotic prophase chromosomes; AW742678; cancer/testis antigen 8; CT8; DlkI; FA1; HOM-TES-14; Ly107; MGC104417; Peg9; pG2; pref-1; SCP1; SCP-1; Sycp1; Synaptonemal complex protein 1; testicular tissue protein Li 191; ZOG
SYCP1
Primary
Antigen affinity chromatography
0.86 mg/mL
Western Blot
Unconjugated
Rabbit
RUO
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q15431
6847
Recombinant protein encompassing a sequence within the C-terminus region of human SCP1. The exact sequence is proprietary.
Antibody

Specifications

SCP1
Polyclonal
Unconjugated
SYCP1
A coiled-coil related protein specific for synapsed regions of meiotic prophase chromosomes; AW742678; cancer/testis antigen 8; CT8; DlkI; FA1; HOM-TES-14; Ly107; MGC104417; Peg9; pG2; pref-1; SCP1; SCP-1; Sycp1; Synaptonemal complex protein 1; testicular tissue protein Li 191; ZOG
Rabbit
Antigen affinity chromatography
RUO
6847
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.86 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q15431
SYCP1
Recombinant protein encompassing a sequence within the C-terminus region of human SCP1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions