missing translation for 'onlineSavingsMsg'
Learn More

Airgas™ Transportable Pure Gases and Mixtures (48L Size) from Restek

Catalog No. 06802417
Encompass
Change view
Click to view available options
Concentration:
100 ppm
1000 ppm
15 ppm
99.80%
CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen
CO 1%, CO2 1%, H2 1%, O2 1% in Nitrogen
CO 1%, CO2 1%, Methane 1%, Ethane 1%, Ethylene 1%, Ecetylene 1% in Nitrogen
Components:
C1-C6 n-Paraffins in Helium
C1-C6 n-Paraffins in Nitrogen
C2-C6 Olefins in Nitrogen
Carbon Dioxide
Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen
Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene, Ecetylene in Nitrogen
Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, n-Butane, Propyne in Nitrogen
n-Butane, Isobutane, cis-2-Butene, trans-2-Butene, 1-Butene, Isobutene, 1,3-Butadiene, Ethyl Acetylene in Nitrogen
Type:
Air Standard
Transportable Air Standard
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Components Type
06802417 99.80% Carbon Dioxide Air Standard
06802470 CO 0.5%, CO2 0.5%, H2 0.5%, O2 0.5% In nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard
06802476 CO 1%, CO2 1%, Methane 1%, Ethane 1%, Ethylene 1%, Ecetylene 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Methane, Ethane, Ethylene, Ecetylene in Nitrogen Transportable Air Standard
06802473 CO 1%, CO2 1%, H2 1%, O2 1% in Nitrogen Carbon Monoxide, Carbon Dioxide, Hydrogen, Oxygen in Nitrogen Transportable Air Standard
06802490 1000 ppm C1-C6 n-Paraffins in Helium Transportable Air Standard
06802493 100 ppm C1-C6 n-Paraffins in Nitrogen Transportable Air Standard
06802497 100 ppm C2-C6 Olefins in Nitrogen Transportable Air Standard
06802504 15 ppm n-Butane, Isobutane, cis-2-Butene, trans-2-Butene, 1-Butene, Isobutene, 1,3-Butadiene, Ethyl Acetylene in Nitrogen Transportable Air Standard
06802502 15 ppm Methane, Ethane, Ethylene, Acetylene, Propane, Propylene, n-Butane, Propyne in Nitrogen Transportable Air Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 06802417 Supplier Chromatographic Supplier No. RK 34452

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for peak identification work

  • CGA 165 connection and can deliver large volumes of sample
  • Scotty™ (Air Liquide) 48
  • Contents: 48L
  • Pressure: 300 psig (21 bar)
  • Outlet Fitting: CGA 165
  • Weight: 1.75 lb. (0.8kg)
  • Dimensions: 4 in. diameter × 16.25 in. height (10.2 × 41cm)
  • DOT Specifications: 39 NRC

All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements

Specifications

Certifications/Compliance DOT 39 NRC
Components Carbon Dioxide
Diameter (Metric) 10.2 cm
Diameter (English) 4 in.
Concentration 99.80%
Gas Type Carbon Dioxide
Volume (Metric) Gas 48 L
For Use With (Application) Ideal for peak identification work
Height (English) 16.25 in.
Height (Metric) 41 cm
Pressure 300 psi
Weight (English) 1.75 lb.
Volume (English) Gas 12.68 gal.
Weight (Metric) 0.8 kg
Type Air Standard
Show More Show Less