missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scopolamine Methyl Bromide 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| UN Number | 1544 |
| Formula Weight | 398.30 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Scopolamine Methyl Bromide |
Safety and Handling
| EINECSNumber | (5)-3899 |
| RTECSNumber | YM3675000 |
| TSCA | Yes |