accessibility menu, dialog, popup

UserName

BrCH3NCH3(S)(S)(R)O(S)OOOH

CAS RN 155-41-9

IUPAC Name: (1S,2R,4S,5S)-7-[(3-hydroxy-2-phenylpropanoyl)oxy]-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide
Synonyms: methylscopolamine bromide
Molecular Weight (g/mol): 398.3
Molecular Formula: C18H24BrNO4
InChi Key: CXYRUNPLKGGUJF-ODDNLWGVSA-M
SMILES: [Br-].C[N+]1(C)[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)C(CO)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results