missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCIN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112705
Description
SCIN Polyclonal Antibody for Western Blot, ICC/IF, ELISA
The SCIN gene, also known as scinderin, encodes for a calcium-dependent actin filament-severing protein that is involved in the regulation of the cytoskeleton. It plays a significant role in actin filament dynamics, influencing cellular processes such as cell shape, motility, secretion, and division. SCIN has been identified as a novel oncogene, with its abnormal expression strongly correlated to various cancers, including glioma and uterine corpus endometrial carcinoma. Specific mutations like S673F and S381Y in SCIN are linked with poor prognosis and are potential targets for cancer diagnosis, prognosis, and treatment strategies.Specifications
| SCIN | |
| Polyclonal | |
| Unconjugated | |
| SCIN | |
| Adseverin; AW545522; Gelsolin-like protein; KIAA1905; SCIN; Scind; Scinderin | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20259, 85477 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 7 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q60604, Q9Y6U3 | |
| SCIN | |
| Recombinant Human Adseverin protein (556-703AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |