missing translation for 'onlineSavingsMsg'
Learn More

SciMatCo™ Stak-a-Cab™ Floor Stand

Keep your important items off the floor by adding the SciMat™ Stak-a-Cab™ Floor Stand to your Stak-A-Cab storage cabinets,

$377.86 - $567.92

Specifications

Description Stak-A-Cab Floor Stand
Product Type Floor Stand
For Use With (Application) Safe stacking of the Stak-a-Cabs up to three units high
Width (Metric) Exterior 118.11 cm
View More Specs

Includes: Four built in adjustable leveling feet

Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
17988486
View Documents Promotion Details
SciMatCo™
SC1461
Blue
Each for $377.86
Only null left
 
17988485
View Documents Promotion Details
SciMatCo™
SC1861
Yellow
Each for $567.92
Only null left
 
17988487
View Documents Promotion Details
SciMatCo™
SC2061
Gray
Each for $377.86
Only null left
 
Description

Description

  • Allows for the safe stacking of the Stak-a-Cabs up to three units high
  • Allows easier and safer access when cabinet is placed on the floor
  • Four built in leveling feet to level cabinets on uneven surfaces
  • Color to blend in with the bottom cabinet All units 46.5W x 4.625H x 17.75 in. D
Specifications

Specifications

Stak-A-Cab Floor Stand
Floor Stand
Safe stacking of the Stak-a-Cabs up to three units high
118.11 cm
Videos
SDS
Documents

Documents