missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) trifluoromethanesulfonate, 98%

CAS: 144026-79-9 | C3F9O9S3Sc | 492.146 g/mol

Supplier:  Thermo Scientific Chemicals 04056603

 View more versions of this product

Catalog No. AA4056603


Only null left
Explore available promotions

Description

Description

Scandium(III) trifluoromethanesulfonate is widely used as a catalyst in hydrothiolation, selective two-electron reduction of oxygen by ferrocene derivatives and vinylogous Fridel-crafts alkylation of indoles and pyrrole in water. It is involved in the Mukaiyama aldol addition and stereochemically catalyzes the radical polymerization of acrylates. It acts as a Lewis acid catalyst and used in the synthesis of bullvalone via a stabilized sulfur ylide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Scandium(III) trifluoromethanesulfonate
Powder
C3F9O9S3Sc
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
HZXJVDYQRYYYOR-UHFFFAOYSA-K
scandium(3+);trifluoromethanesulfonate
2734571
98%
144026-79-9
1 g
MFCD00798539
Soluble in water,alcohol and acetonitrile.
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
492.146
492.16
Hygroscopic

Chemical Identifiers

144026-79-9
492.146
HZXJVDYQRYYYOR-UHFFFAOYSA-K
2734571
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
C3F9O9S3Sc
MFCD00798539
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
scandium(3+);trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only