accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOFFFSOOOSc

CAS RN 144026-79-9

IUPAC Name: scandium(3+) tritrifluoromethanesulfonate
Synonyms: scandium(III) trifluoromethanesulfonate
Molecular Weight (g/mol): 492.15
Molecular Formula: C3F9O9S3Sc
InChi Key: HZXJVDYQRYYYOR-UHFFFAOYSA-K
SMILES: [Sc+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)