missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) bis(trifluoromethylsulfonyl)imide

Catalog No. p-7080239
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46948MD 250 mg
AA4694803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA46948MD Supplier Thermo Scientific Chemicals Supplier No. 046948MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 176726-07-1 | C6F18N3O12S6Sc | 885.362 g/mol

Scandium(III) bis(trifluoromethylsulfonyl)imide is used as Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 176726-07-1
Molecular Formula C6F18N3O12S6Sc
Molecular Weight (g/mol) 885.362
MDL Number MFCD03427000
InChI Key FUXLYEZEIZAKTL-UHFFFAOYSA-N
Synonym scandium iii trifluoromethanesulfonimide
PubChem CID 131875098
IUPAC Name bis(trifluoromethylsulfonyl)azanide;scandium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sc+3]

Specifications

Melting Point 199°C to 204°C
Color White to Yellow
Linear Formula [(CF3SO2)2N]3Sc
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 885.37
Physical Form Solid
Chemical Name or Material Scandium (III) bis (trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only