missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) 2,4-pentanedionate hydrate

CAS: 699012-88-9 | C15H21O6Sc | 342.28 g/mol

$254.21 - $985.04

Chemical Identifiers

CAS 699012-88-9
Molecular Formula C15H21O6Sc
Molecular Weight (g/mol) 342.28
MDL Number MFCD00064715
InChI Key NYPXMGRKLWIPFL-UHFFFAOYSA-N
Synonym tris 4-hydroxypent-3-en-2-one hydrate scandium
PubChem CID 119030428
IUPAC Name (Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate
SMILES [Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4444803
View Documents
Thermo Scientific Chemicals
04444803
1 g
Each for $254.21
Only null left
 
AA4444806
View Documents
Thermo Scientific Chemicals
04444806
5 g
Each for $985.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

699012-88-9
342.28
NYPXMGRKLWIPFL-UHFFFAOYSA-N
119030428
[Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C15H21O6Sc
MFCD00064715
tris 4-hydroxypent-3-en-2-one hydrate scandium
(Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate

Specifications

174°C to 186°C
699012-88-9
1 g
MFCD00064715
NYPXMGRKLWIPFL-UHFFFAOYSA-N
(Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate
119030428
Odorless
Colorless
Powder
C15H21O6Sc
tris 4-hydroxypent-3-en-2-one hydrate scandium
[Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
342.28
342.26 (Anhydrous)
Scandium(III) 2, 4-pentanedionate hydrate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-194-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only