accessibility menu, dialog, popup

UserName

OOOOOOCHHCCHScH3CH3CCH3H3CCH3H3C

CAS RN 699012-88-9

IUPAC Name: scandium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: scandium(3+) triacetylacetonate
Molecular Weight (g/mol): 342.28
Molecular Formula: C15H21O6Sc
InChi Key: NYPXMGRKLWIPFL-UHFFFAOYSA-N
SMILES: [Sc+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results