missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP5 Monoclonal Antibody (K06322_12G11), Invitrogen™
Mouse Monoclonal Antibody
$1693.48 - $2847.38
Specifications
| Antigen | SCAMP5 |
|---|---|
| Clone | K06322_12G11 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: LPKFIPLKPC FYQDFEADIP
SCAMP5 Monoclonal antibody specifically detects SCAMP5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| SCAMP5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| hSCAMP5; Sc5; scamp5; scamp5a; Secretory carrier membrane protein 5; secretory carrier membrane protein 5a; secretory carrier membrane protein 5-like; secretory carrier-associated membrane protein 5; wu:fc30b07; zgc:77438 | |
| Human SCAMP5 peptide (immunogen range is 11-30). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SCAMP5 |
| K06322_12G11 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q8TAC9, Q9JKD3 | |
| 192683, 56807 | |
| IgG1 | |
| Protein A | |
| Antibody |