missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SCAMP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1734
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1734 Supplier Invitrogen™ Supplier No. PA1734
Only null left

Rabbit Polyclonal Antibody

PA1-734 detects secretory carrier membrane protein 2 (SCAMP 2) from mouse testes and AtT20 cell extract as well as Chinese hamster ovary (CHO) cell extract. PA1-734 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~38 kDa protein representing SCAMP 2 from CHO cell extract. PA1-734 immunizing peptide corresponds to amino acid residues 1-17 from human SCAMP 2. This sequence is completely conserved between human and mouse. PA1-734 immunizing peptide (Cat. # PEP-151) is available for use in neutralization and control experiments.

Secretory carrier membrane proteins (SCAMPs) are proteins that are components of post-Golgi membranes. These proteins are implicated to function in membrane trafficking. In fibroblasts, SCAMPs are concentrated in compartments that are involved in the recycling of cell surface receptors and endocytosis. In neurons, SCAMPs are associated with synaptic vesicles, secretion granules and transporter vesicles. SCAMPs are composed of four central transmembrane regions and a cytoplasmic tail. Of the five known SCAMPs, SCAMPs 1-3 contain cytoplasmic N-terminal regions with NPF repeats. NPF repeats are found to interact with EH domain proteins that function in budding of transport vesicles from the plasma membrane or the Golgi complex. SCAMPs 4 and 5 lack the N-terminal NPF repeats. SCAMPs 1-4 are all ubiquitously coexpressed while SCAMP 5 is only detectable in the brain. Studies show that SCAMP 5 is expressed late in development which is coincident with expansion of mature synapses.

Specifications

Antigen SCAMP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Scamp2
Gene Accession No. O15127, Q9ERN0
Gene Alias AI875466; I79_023583; Sc2; scamp 2; SCAMP2; Secretory carrier membrane protein 2; secretory carrier-associated membrane protein 2; testis secretory sperm-binding protein Li 219p
Gene Symbols Scamp2
Host Species Rabbit
Immunogen Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10066, 100756209, 24044
Target Species Human, Mouse, Hamster
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less