missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals SB 203580, 98%
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | CAS: 152121-47-6 | C21H16FN3OS, C21H16FN3OS | 377.44 g/mol
$293.51 - $733.75
Chemical Identifiers
| CAS | 152121-47-6 |
|---|---|
| Molecular Formula | C21H16FN3OS |
| Synonym | 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ61482MB
|
Thermo Scientific Chemicals
J61482MB |
25 mg |
Each for $293.51
|
|
|||||
|
AAJ61482MC
|
Thermo Scientific Chemicals
J61482MC |
100 mg |
Each for $733.75
|
|
|||||
|
AAJ61482MF
|
Thermo Scientific Chemicals
J61482MF |
50 mg |
Each for $418.78
|
|
|||||
Description
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 152121-47-6 | |
| C21H16FN3OS | |
| 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole |
Specifications
| White to Off-white | |
| Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | |
| C21H16FN3OS | |
| 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole | |
| Signal Transduction Reagent | |
| CDMGBJANTYXAIV-UHFFFAOYNA-N | |
| 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine | |
| 98% |
| -30°C to -10°C | |
| 152121-47-6 | |
| 377.44g/mol | |
| 235°C to 247°C | |
| Powder | |
| CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1 | |
| 377.44 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only