missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SB 203580, 98%

Catalog No. AAJ61482MB
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61482MB 25 mg
AAJ61482MC 100 mg
AAJ61482MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61482MB Supplier Thermo Scientific Chemicals Supplier No. J61482MB

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | CAS: 152121-47-6 | C21H16FN3OS, C21H16FN3OS | 377.44 g/mol

Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152121-47-6
Molecular Formula C21H16FN3OS
Synonym 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole

Specifications

Quantity 25 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
Formula Weight 377.44g/mol
Melting Point 235°C to 247°C
Product Type Signal Transduction Reagent
Form Powder
InChI Key CDMGBJANTYXAIV-UHFFFAOYNA-N
SMILES CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
IUPAC Name 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine
Molecular Weight (g/mol) 377.44
Assay 98%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only