Learn More
Thermo Scientific Chemicals SB 203580, 98%
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | CAS: 152121-47-6 | C21H16FN3OS, C21H16FN3OS | 377.44 g/mol
Supplier: Thermo Scientific Chemicals J61482MC
Description
Highly specific new inhibitor of p38 kinase and MAP Kinase homologues
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 100 mg | |
| -30°C to -10°C | |
| 98% | |
| C21H16FN3OS | |
| 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole | |
| Signal Transduction Reagent | |
| CDMGBJANTYXAIV-UHFFFAOYNA-N | |
| 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine |
| White to Off-white | |
| Highly specific new inhibitor of p38 kinase and MAP Kinase homologues | |
| 152121-47-6 | |
| 377.44g/mol | |
| 235°C to 247°C | |
| Powder | |
| CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1 | |
| 377.44 |
Chemical Identifiers
| 152121-47-6 | |
| C21H16FN3OS | |
| 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)imidazole |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only