Learn More
Sartorius Reusable Sterility Testing System: Silicone Adapter
Silicone Adapter for use with Reusable Sterility Testing System.
Supplier: Sartorius 16968
Description
- With stoppers drilled to fit filter holder base
- Allows sterile filtration system to be used with Sartorius and other stainless-steel and PVC manifolds
Specifications
| Reusable | |
| Silicone | |
| Silicone Adapter |
| Reusable Sterility Testing System | |
| Non-sterile |