missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Minisart™ NY Syringe Filters: Nonsterile

For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convenience.

Manufacturer:  Sartorius 1784BK

 View more versions of this product

Catalog No. 14-555-346



Description

Description

For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convenience.

Ready-to-use units, which offer flow rates at low inlet pressures, make rapid filtration possible. Fitted on a standard syringe.

  • Particle removal
  • Easy to handle
  • Proven quality
  • 100% tested during manufacture

Specifications

Specifications

25 mm
For rapid small volume filtration of both aqueous and solvent-based samples with maximum user convinience
500/Pk.
Polyamide (Nylon) Filter, Glass Fiber Prefilter
Non-sterile
Yes
Luer Slip
Syringe Filter
0.2 μm
SDS
Documents
Promotions

Promotions