missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ Balance Display and Control Units
GREENER_CHOICE

Catalog No. 14557557
Change view
Click to view available options
Includes:
Backlit black and white graphic display and tactil
Backlit LCD and tactile keys
Color TFT graphic display and touch screen
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
14557557 Backlit black and white graphic display and tactil
14557556 Backlit LCD and tactile keys
14557558 Color TFT graphic display and touch screen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14557557 Supplier Sartorius Supplier No. YAC01MSU

Variety of displays and control units available

For use with Cubis™ balances.

Specifications

For Use With (Equipment) Cubis Balance
Includes Backlit black and white graphic display and tactil
Type Control Unit