missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Balance Weighing Hooks
GREENER_CHOICE

Catalog No. 14557565
Change view
Click to view available options
Weight (Metric):
0.9 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
14557565 0.9 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14557565 Supplier Sartorius Supplier No. 69EA0040

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Expand the uses of your balance with the below-balance weighing hook.

  • For high capacity models (12kg and above)
  • For use on CPA Balances, Extend ED Balances, Master Pro LA Balances, SE and ME Ultramicro and Microbalances, M-Power Balances

Specifications

For Use With (Equipment) 12 kg and above CPA Balance, Master Pro LA Balance
Weight (English) 2 lb.
Weight (Metric) 0.9 kg
Type Weighing Hook