Learn More
Sartorius Balance Density Determination Kits
GREENER_CHOICE
Catalog No. 14557579
Change view
Click to view available options
For Use With (Equipment):
CPA Balance
Cubis Balance
Extend ED, LA, LP Precision Balance
Product Line:
Cubis I
Cubis I, 1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Line
14557579 Extend ED, LA, LP Precision Balance -
14557545 Cubis Balance Cubis I
14557546 Cubis Balance Cubis I, 1 mg
09368148 CPA Balance -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14557579 Supplier Sartorius Supplier No. YDK01LP
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use with Sartorius balances to determine the density of solids and liquids
All kits include
  • 2 Beakers (7mm and 55mm dia.)
  • Bar frame
  • Thermometer with retainer clip
  • Glass plummet
  • Sieve for immersing samples (pan hanger assembly)
  • Sample holder (pan hanger assembly)
  • Metal platform
CPA Kit includes
  • Gasket for ME models
  • Compensating disk for ME235S/P, ME254S models
  • 3 adapters
Cubis Kits include
  • Adapter for LA/LP models with analytical draft shield
Extended ED Kit includes
  • Adapters for LA/LP models: 1 with and 1 without analytical draft shield
  • Compensating disk for LA/LP2300D models
Specifications
For Use With (Equipment) Extend ED, LA, LP Precision Balance
For Use With (Application) Determining the density of solids and liquids
Includes 2 Beakers (7 mm and 55 mm dia.), Bar Frame, Thermome
Type Density Determination Kit