Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
$24.54 - $366.47
Specifications
Disposable Reusable
For Use With (Equipment) Sartorius Biosart 100
Sterility Non-sterile
View More Specs
Products 4
Catalog Number Mfr. No. Type Material For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Type Material For Use With (Application) Price Quantity  
14-555-780
View Documents
Sartorius
16417
Adapter ABS Copolymer For easy transfer of the membrane onto agar
Each for $24.54
Only null left
14-555-778
View Documents
Sartorius
16414
Adapter Silicone Fits Biosart 100 Monitor onto 16840 or 16841 base
Each for $33.46
Only null left
14-555-781
View Documents Promotion Details
Sartorius
16835
Adapter Stainless Steel Fits Biosart 100 Monitors onto Combisart manifolds 16844, 16842 and 16843
Each for $366.47
Only null left
14-555-817
View Documents
Sartorius
00279
Stopper Silicone Replacement for 16835
Each for $50.98
Only null left
Description
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780
  • Biosart 100 Membrane Lifter
  • For easy transfer of the membrane onto agar
Adapter, Polypropylene - 14-555-779
  • Biosart 100 Adapter
  • Fits Biosart 100 Monitor onto 50 mm supports
Adapter, Silicone - 14-555-778
  • Biosart 100 Adapter
  • 50 mm stainless-steel frit and silicone gasket
  • Fits Biosart 100 Monitor onto 16840 or 16841 base
Adapter, Stainless Steel - 14-555-781
  • Biosart 100 Adapter to accommodate various funnel types
  • Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
Stopper, Silicone - 14-555-817
  • Replacement stopper for 16385
  • 16385 adapts Biosart 100 Monitors on Combisart manifold
Specifications
Specifications
Reusable
Sartorius Biosart 100
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'