missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle

Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.
Supplier: Sartorius 18024E
Description
- The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size
Chemical Identifiers
| Single-use | |
| Polystyrene | |
| Sterile |
| Sartolab | |
| Polystyrene |
Specifications
| Single-use | |
| Polystyrene | |
| 500 mL |
| Sartolab | |
| Polystyrene | |
| Sterile |