missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartolab™ Single-Use Polystyrene Storage Bottle
GREENER_CHOICE

Catalog No. 14578811
Change view
Click to view available options
Sample Volume:
150 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
14578811 150 mL
14578810 250 mL
14578812 500 mL
14578813 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14578811 Supplier Sartorius Supplier No. 180222

Sartorius Sartolab single-use polystyrene bottles for storage of media, buffers, and other aqueous solutions.

  • The bottles can be used with all Sartolab™ RF | BT vacuum filtration units with a 45 mm neck size

Specifications

Disposable Single-use
For Use With (Equipment) Sartolab
Material Polystyrene
Material (Membrane) Polystyrene
Sample Volume 150 mL
Sterility Sterile