missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartobind™ Rapid A Lab

Catalog No. p-200113465
Change view
Click to view available options
Quantity:
1/Ea.
4/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14222462 1/Ea.
14222463 4/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222462 Supplier Sartorius Supplier No. SBLRA025EL1
Only null left

Rapid A Lab Affinity Purification Unit with high flow rates.

  • High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes
  • Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity
  • Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration
  • Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system
  • Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production

Specifications

Flow Rate 2.5 to 25 mL/min.
Column Type Affinity
Stationary Phase Agarose or Protein A
Diameter (Metric) Inner Column 25 mm
Volume (Metric) 0.5 mL
Quantity 1/Ea.