missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartobind Lab IEX Q75
Description
- Process-Ready Sartobind™ Membrane Adsorbers Sartobind™ Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format.
- They are based on the patented macroporous Sartobind™ membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a media chromatography matrix with superior flow rates and high binding capacity.
- Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity. In addition, these units utilize the same membrane adsorber platform as found in Sartobind™ capsules and cassettes, making them ideal for research and development work, prior to convenient scale-up for manufacturing.
- Ultimate Versatility Sartobind™ Lab units for ion exchange are available with strong or weak anion (Q and D) and strong cation (S) exchangers, and in three bed volumes, ranging from 0.41 to 2.8 mL, corresponding to dynamic binding capacities between 12 and 80 mg protein per unit.
- Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used 1,000 times, ensuring high-cost efficiency.
- Greatly reduced cycle times
- Achieve higher yields in less time
- Purify equipment-free or with an FPLC system
Specifications
Specifications
| Bed Volume | 2.8 mL |
| Connectors | Luer Lock |
| Dead Volume | 5.7 mL |
| Disposable | Reusable |
| Filter Type | Air Syringe Filter |
| For Use With (Equipment) | Syringe |
| Material | Polyester |
| Material (Membrane) | Cellulose |
| Pore Size | 5 μm |
| Sterility | Non-sterile |
| Show More |