missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Sartobind Lab Affinity Pr A
Description
- High-Speed Purification Sartobind™ Lab Protein A affinity membrane adsorbers are ready-to-use units in a syringe filter format.
- The macorporous matrix makes them the ideal tool for rapid purification of immunoglobulins (IgG) and monoclonal antibodies (mAbs) by providing superior flow rates and reduced cycle times when compared to conventional columns.
- A complete purification cycle, whether by syringe or on an FPLC system, takes less than 5 minutes.
- Ultimate Versatility Sartobind™ Lab Protein A units are available with a 2 mL bed volume, providing binding capacities of 10 to 15 mg IgG per unit. Equipped with standard Luer lock connectors, Sartobind™ Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications.
- The units are compatible with most common protein A purification buffers and can be regenerated for multiple purification cycles, ensuring high-cost efficiency.
- Greatly reduced cycle times
- Achieve higher yields in less time
- Purify equipment-free or with an FPLC system
- Specifications:
- Ligand: Protein A
- Reference Protein: Human polyclonal IgG
Specifications
Specifications
| Bed Volume | 2.1 mL |
| Disposable | Reusable |
| Filter Type | Air Syringe Filter |
| For Use With (Equipment) | Syringe |
| Material | Polyester |
| Material (Membrane) | Cellulose |
| Pore Size | 0.45 μm |
| Sterility | Non-sterile |
| For Use With (Application) | mAb Purification |
| Membrane Type | Affinity |
| Show More |