missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV/SARS-CoV-2 Spike Protein S1 Chimeric Recombinant Mouse Monoclonal Antibody (CR3022)
GREENER_CHOICE

Catalog No. MA551599
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551599 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551599 Supplier Invitrogen™ Supplier No. MA551599
Only null left

Mouse Recombinant Monoclonal Antibody

This antibody binds the amino acids 318-510 in the S1 domain of the SARS-CoV Spike protein as well as SARS-CoV-2 (COVID-19) Spike protein. The antibody also binds to P462L-substituted S318-510 fragments of the SARS spike protein.

The SARS-CoV-2 virus, responsible for the COVID-19 pandemic, has a spike protein composed of two subunits, S1 and S2. The S1 subunit binds to the human ACE2 receptor, initiating the virus's entry into the cell and recognizing the host cell for the fusion process. The SARS spike protein S1, responsible for binding to the ACE2 receptor during the 2002-2003 SARS outbreak, shares a high degree of similarity with the SARS-CoV-2 spike protein S1, but there are differences in their amino acid sequence that may affect their binding affinity to the ACE2 receptor. Both spike proteins have a trimeric structure, with each subunit consisting of two domains, the N-terminal domain and the receptor-binding domain. The RBD is the part of the S1 subunit that binds to the ACE2 receptor, while the NTD is involved in the recognition of other host factors.

Specifications

Antigen SARS-CoV/SARS-CoV-2 Spike Protein S1 Chimeric
Applications ELISA, Neutralization, Immunocytochemistry, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone CR3022
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Host Species Mouse
Immunogen The original monoclonal antibody was generated through an scFv library derived from a peripheral blood lymphocytes of a patient exposed to the SARS-CoV.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less