missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV/SARS-CoV-2 NP Chimeric Recombinant Human Monoclonal Antibody (CR3018 (03-018))
GREENER_CHOICE

Catalog No. MA551593
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551593 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551593 Supplier Invitrogen™ Supplier No. MA551593
Only null left

Human Recombinant Monoclonal Antibody

This antibody binds the amino acid residues between 11-19 of the N protein of the SARS CoV as well as SARS-CoV-2 (COVID-19) nucleocapsid protein.

Human coronaviruses are the major cause of upper respiratory tract illness in humans, such as the common cold. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The most prominent protein in the culture supernatants infected with SARS virus is a 46 kDa nucleocapsid protein. This suggests that the nucleocapsid protein is a major immunogen that may be useful for early diagnostics. SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus.

Specifications

Antigen SARS-CoV/SARS-CoV-2 NP Chimeric
Applications ELISA, Immunocytochemistry
Classification Recombinant Monoclonal
Clone CR3018 (03-018)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias nucleoprotein
Host Species Human
Immunogen Whole irradiated virion.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less