Learn More
Invitrogen™ SARS-CoV-2 Spike Protein (RBD) Recombinant Human Monoclonal Antibody (P05DHu), Alexa Fluor™ 488, eBioscience™
Description
Description: This antibody recognizes the RBD domain of SARS CoV2 Spike protein. Applications Tested: This P05DHu Alexa Fluor™ 488 conjugate has been tested by flow cytometric analysis of HEK 293 cells transfected with a vector encoding full length spike protein from SARS CoV2. This may be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | SARS-CoV-2 Spike Protein (RBD) |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | P05DHu |
| Concentration | 0.5 mg/mL |
| Conjugate | Alexa Fluor 488 |
| Formulation | PBS, proprietary buffer with 0.09% sodium azide; pH 7.2-7.4 |
| Gene Alias | Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 |
| Host Species | Human |
| Immunogen | This antibody was developed using computationally derived variants of SARS-CoV antibody structures and is shown to bind Spike Protein S1 subunit (RBD) of SARS CoV2. |
| Show More |