missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein (RBD) Chimeric Recombinant Rabbit Monoclonal Antibody (Sb#45)
GREENER_CHOICE

Catalog No. MA551649
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551649 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551649 Supplier Invitrogen™ Supplier No. MA551649
Only null left

Rabbit Recombinant Monoclonal Antibody

This antibody recognizes and binds the SARS CoV 2 C-terminal receptor binding domain (RBD) located in the extracellular portion of the spike protein.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.

Specifications

Antigen SARS-CoV-2 Spike Protein (RBD) Chimeric
Applications ELISA, Functional Assay, Inhibition Assays
Classification Recombinant Monoclonal
Clone Sb#45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Host Species Rabbit
Immunogen This clone was originally isolated in a form of a synthetic nanobody (sybody) via a 'target swap' selection procedure against RBD-vYFP using ribosomal display and against RBD-Fc fusion during phage display rounds.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG Fc
Show More Show Less