missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (84C4A), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

$993.31 - $2315.17

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Clone 84C4A
Concentration 0.5 mg/mL
Applications ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIMA537455
View Documents
Invitrogen™
MA537455
20 μL
Each for $993.31
Only null left
 
PIMA537207
View Documents
Invitrogen™
MA537207
200 μL
Each for $2,315.17
Only null left
 
Description

Description

SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
Specifications

Specifications

SARS-CoV-2 Nucleocapsid
0.5 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 0.02% sodium azide
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
N
IgG κ
Protein A
Antibody
84C4A
ELISA
Unconjugated
Rabbit
Virus
P0DTC9
43740575
SARS-CoV-2 nucleoprotein (NP) specific peptide.
Primary
4°C
N
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'