missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (75G5a), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
$947.12 - $2212.50
Specifications
| Antigen | SARS-CoV-2 Nucleocapsid |
|---|---|
| Clone | 75G5a |
| Concentration | 0.5 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
PIMA537453
|
Invitrogen™
MA537453 |
20 μL |
Each for $947.12
|
|
|||||
|
PIMA537205
|
Invitrogen™
MA537205 |
200 μL |
Each for $2,212.50
|
|
|||||
Description
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.Specifications
| SARS-CoV-2 Nucleocapsid | |
| 0.5 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.02% sodium azide | |
| COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
| N | |
| IgG κ | |
| Protein A | |
| Antibody |
| 75G5a | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| P0DTC9 | |
| 43740575 | |
| SARS-CoV-2 nucleoprotein (NP) specific peptide. | |
| Primary | |
| 4°C | |
| N |