missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Neutralizing Ab ELISA Kit

Catalog No. BMS2326TEN
Encompass
Change view
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
5 x 96 Tests
96 Tests
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2326TEN 10 x 96 Tests
BMS2326 96 Tests
BMS2326TWO 2 x 96 Tests
BMS2326FIVE 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BMS2326TEN Supplier Invitrogen™ Supplier No. BMS2326TEN
Only null left

ELISA

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.

Specifications

Content And Storage 2°C to 8°C
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Host Species Human
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Control, SA-HRP Conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
Protein SARS-CoV-2 Spike Protein RBD
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 5 μL, Serum 5 μL
Shipping Condition Wet Ice
Target SARS-CoV-2 Spike Protein RBD
Test Time 40 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less