missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sarpogrelate hydrochloride, 98%
Supplier: Thermo Scientific™ 461910010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Sarpogrelate hydrochloride | |
| 97.5% min. (HPLC) | |
| C24H31NO6·HCl | |
| Solubility in water: soluble. Other solubilities: soluble in dimethylsulfoxide and methanol | |
| 465.97 | |
| 465.97 |
| 135159-51-2 | |
| Glass Bottle | |
| 1g | |
| POQBIDFFYCYHOB-UHFFFAOYNA-N | |
| 444005 | |
| 98% |
Chemical Identifiers
| 135159-51-2 | |
| 465.97 | |
| 444005 |
| C24H31NO6·HCl | |
| POQBIDFFYCYHOB-UHFFFAOYNA-N |
Safety and Handling
GHS H Statement
Very toxic to aquatic life.
GHS Signal Word: Warning
RUO â Research Use Only